Ethyl-2-methylnicotinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0860-25G E0860-5G
TCE0860-25GEA 317.58 CAD
TCE0860-25G TCE0860-5G
Ethyl-2-methylnicotinate ≥98.0% (by GC, titration analysis)
Ethyl 2-methylnicotinate

CAS Number: 1721-26-2
MDL Number: MFCD00006336
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 103
Specific Gravity (20/20): 1.08
Lambda max.: 268 nm (EtOH)


Other Product Names / Synonyms: 2-Methylnicotinic Acid Ethyl Ester, Ethyl 2-Methylpyridine-3-carboxylate, 2-Methylpyridine-3-carboxylic Acid Ethyl Ester

Formula: C₉H₁₁NO₂
Boiling Pt: 125 °C
Density: 1.08 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00006336
CAS Number: 1721-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1e878a8d702899b56c67915c19506b21c59a93a&vid=a1e878a8d702899b56c67915c19506b21c59a93a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR