Ethyl-1H-tetrazole-5-acetate ≥98.0%

Supplier: TCI America

Synonyms: 1H-Tetrazole-5-acetic Acid Ethyl Ester, Ethyl 2-(2H-tetrazol-5-yl)acetate

E0859-25G E0859-5G
TCE0859-25GEA 418.7 CAD
TCE0859-25G TCE0859-5G
Ethyl-1H-tetrazole-5-acetate ≥98.0%
Ethyl-1H-tetrazole-5-acetate

CAS Number: 13616-37-0
MDL Number: MFCD00274213
Molecular Formula: C5H8N4O2
Molecular Weight: 156.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 128


Other Product Names / Synonyms: 1H-Tetrazole-5-acetic Acid Ethyl Ester

Formula: C₅H₈N₄O₂
Melting Pt: 128 °C
MDL Number: MFCD00274213
CAS Number: 13616-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89db68080a187a5711006a0945aaf4890eb2fb22&vid=89db68080a187a5711006a0945aaf4890eb2fb22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR