Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0855-1G E0855-5G
TCE0855-1GEA 52.93 CAD
TCE0855-1G TCE0855-5G
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride

CAS Number: 52763-21-0
MDL Number: MFCD00012792
Molecular Formula: C15H19NO3
Molecular Weight: 297.78
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)

Formula: C₁₅H₂₀ClNO₃
Melting Pt: 165 °C
MDL Number: MFCD00012792
CAS Number: 52763-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3b6f4c8c02090bff4ff2620fa72370622243056&vid=c3b6f4c8c02090bff4ff2620fa72370622243056&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR