Ethyl-6-chloronicotinate ≥97.0%

Supplier: TCI America

Synonyms: Ethyl 6-chloronicotinate

E0852-25G E0852-5G
TCE0852-25GEA 146.94 CAD
TCE0852-25G TCE0852-5G
Ethyl-6-chloronicotinate ≥97.0%
Ethyl 6-chloronicotinate

CAS Number: 49608-01-7
MDL Number: MFCD00082739
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 1.24


Other Product Names / Synonyms: 6-Chloronicotinic Acid Ethyl Ester, Ethyl 2-Chloro-5-pyridinecarboxylate, 2-Chloro-5-pyridinecarboxylic Acid Ethyl Ester

Formula: C₈H₈ClNO₂
MW: 171.58
Boiling Pt: 150 °C
Density: 1.24 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00082739
CAS Number: 49608-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=918421d169e65562de5f52f4e38ffb287f86fb97&vid=918421d169e65562de5f52f4e38ffb287f86fb97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR