Ethyl-2-(p-Toluenesulfonyloxy)acetate ≥98.0%

Supplier: TCI America

Synonyms: 2-(p-Toluenesulfonyloxy)acetic Acid Ethyl Ester, 2-(p-Toluenesulphonyloxy)acetic Acid Ethyl Ester

E0851-1G E0851-5G
TCE0851-1GEA 297.04 CAD
TCE0851-1G TCE0851-5G
Ethyl-2-(p-Toluenesulfonyloxy)acetate ≥98.0%
Ethyl-2-(p-Toluenesulfonyloxy)acetate

CAS Number: 39794-75-7
Molecular Formula: C11H14O5S
Molecular Weight: 258.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 2-(p-Toluenesulfonyloxy)acetic Acid Ethyl Ester

Formula: C₁₁H₁₄O₅S
MW: 258.29 g/mol
Melting Pt: 47 °C
CAS Number: 39794-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ded456aaab2c54c7a4f2b704f85ec93666add6d&vid=5ded456aaab2c54c7a4f2b704f85ec93666add6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR