3-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
E0848-1G
E0848-5G
TCE0848-1GEA
31.6
CAD
TCE0848-1G
TCE0848-5G
3-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Ethoxycarbonyl)phenylboronic acid
CAS Number: 4334-87-6
MDL Number: MFCD02179444
Molecular Formula: C9H11BO4
Molecular Weight: 193.99
Form: Crystal
Color: White
Melting point (°C): 138
Other Product Names / Synonyms: 3-(Ethoxycarbonyl)benzeneboronic Acid
Formula:
C₉H₁₁BO₄ Melting Pt: 138 °C |
MDL Number:
MFCD02179444 CAS Number: 4334-87-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...