Ethyl cyano(hydroxyimino)acetate ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

E0847-25G E0847-100G
TCE0847-25GEA 45.46 CAD
TCE0847-25G TCE0847-100G
Ethyl cyano(hydroxyimino)acetate ≥98.0% (by GC, total nitrogen)
Ethyl cyano(hydroxyimino)acetate

CAS Number: 3849-21-6
MDL Number: MFCD00002112
Molecular Formula: C5H6N2O3
Molecular Weight: 142.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: Cyano(hydroxyimino)acetic Acid Ethyl Ester, Ethyl Cyanoglyoxalate Oxime, Oxyma

Formula: C₅H₆N₂O₃
Melting Pt: 133 °C
MDL Number: MFCD00002112
CAS Number: 3849-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4189b446fc9661c298dbe93516cf4a733dea6119&vid=4189b446fc9661c298dbe93516cf4a733dea6119&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR