Ethyl-2-chloronicotinate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

E0844-100G E0844-25G
TCE0844-100GEA 209 CAD
TCE0844-100G TCE0844-25G
Ethyl-2-chloronicotinate ≥98.0% (by GC)
Ethyl 2-chloronicotinate

CAS Number: 1452-94-4
MDL Number: MFCD00674129
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Specific Gravity (20/20): 1.25


Other Product Names / Synonyms: 2-Chloronicotinic Acid Ethyl Ester, Ethyl 2-Chloro-3-pyridinecarboxylate, 2-Chloro-3-pyridinecarboxylic Acid Ethyl Ester

Formula: C₈H₈ClNO₂
MW: 185.61
Boiling Pt: 78 °C
Density: 1.25 (20 °C)
MDL Number: MFCD00674129
CAS Number: 1452-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae69b6cfb9545b401121f95d35f14cd7c98de76b&vid=ae69b6cfb9545b401121f95d35f14cd7c98de76b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR