Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 3,5-dimethyl-2-pyrrolecarboxylate

E0843-25G E0843-5G
TCE0843-25GEA 524.56 CAD
TCE0843-25G TCE0843-5G
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate

CAS Number: 2199-44-2
MDL Number: MFCD00010645
Molecular Formula: C9H13NO2
Molecular Weight: 167.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 272
Melting point (°C): 124


Other Product Names / Synonyms: 3,5-Dimethyl-2-pyrrolecarboxylic Acid Ethyl Ester

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 272 °C
Melting Pt: 124 °C
MDL Number: MFCD00010645
CAS Number: 2199-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=939bd7ba32294fc2a542f9e12f8b7c8095fc1b68&vid=939bd7ba32294fc2a542f9e12f8b7c8095fc1b68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR