Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥97.0% (by GC)

Supplier: TCI America

E0841-25G E0841-5G
TCE0841-25GEA 157.05 CAD
TCE0841-25G TCE0841-5G
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥97.0% (by GC)
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate

CAS Number: 5909-24-0
MDL Number: MFCD00006085
Molecular Formula: C8H9ClN2O2S
Molecular Weight: 232.68
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 62


Other Product Names / Synonyms: 4-Chloro-2-(methylthio)pyrimidine-5-carboxylic Acid Ethyl Ester

Formula: C₈H₉ClN₂O₂S
Boiling Pt: 132 °C
Melting Pt: 62 °C
MDL Number: MFCD00006085
CAS Number: 5909-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=302ed8572e147f4b5c15c8da08a095edd034aae6&vid=302ed8572e147f4b5c15c8da08a095edd034aae6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR