3-Ethoxy-4-methoxybenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

E0835-25G E0835-5G
TCE0835-25GEA 152 CAD
TCE0835-25G TCE0835-5G
3-Ethoxy-4-methoxybenzaldehyde ≥98.0% (by GC)
3-Ethoxy-4-methoxybenzaldehyde

CAS Number: 1131-52-8
MDL Number: MFCD00010128
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 50
Flash Point (°C): 113

Formula: C₁₀H₁₂O₃
Boiling Pt: 155 °C
Melting Pt: 50 °C
Flash Pt: 113 °C
MDL Number: MFCD00010128
CAS Number: 1131-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8df1b8841cd5d0847ea1a270f1435d0a12976dda&vid=8df1b8841cd5d0847ea1a270f1435d0a12976dda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR