N-Ethyl-o-crotonotoluidide ≥95.0%

Supplier: TCI America

Synonyms: (E)-N-Ethyl-N-(o-tolyl)but-2-enamide, N-Crotonyl-N-ethyl-o-toluidine, trans-N-Ethyl-N-(o-tolyl)-2-butenamide

E0814-25G E0814-5G
TCE0814-25GEA 481.9 CAD
TCE0814-25G TCE0814-5G
N-Ethyl-o-crotonotoluidide ≥95.0%
N-Ethyl-o-crotonotoluidide

CAS Number: 124236-29-9
MDL Number: MFCD00026989
Molecular Formula: C13H17NO
Molecular Weight: 203.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: trans-N-Ethyl-N-(o-tolyl)-2-butenamide, N-Crotonyl-N-ethyl-o-toluidine

Formula: C₁₃H₁₇NO
Boiling Pt: 155 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00026989
CAS Number: 124236-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55d9afce70d0067aa327fde76131b594d691bd3f&vid=55d9afce70d0067aa327fde76131b594d691bd3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR