Ethyl-3-amino-3-methylbutanoate hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
E0809-1G
E0809-5G
TCE0809-1GEA
82.63
CAD
TCE0809-1G
TCE0809-5G
Ethyl-3-amino-3-methylbutanoate hydrochloride ≥98.0% (by total nitrogen basis)
Ethyl-3-amino-3-methylbutanoate hydrochloride
CAS Number: 85532-40-7
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 101
Other Product Names / Synonyms: 3-Amino-3-methylbutyric Acid Ethyl Ester Hydrochloride
Formula:
C₇H₁₆ClNO₂ Melting Pt: 101 °C |
MDL Number:
MFCD11844787 CAS Number: 85532-40-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...