3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

E0743-1G E0743-5G
TCE0743-1GEA 155.9 CAD
TCE0743-1G TCE0743-5G
3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid ≥95.0% (by GC, titration analysis)
3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid

CAS Number: 18361-03-0
MDL Number: MFCD00966477
Molecular Formula: C8H6O6S
Molecular Weight: 230.19
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal


Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents, Monomers
Other Product Names / Synonyms: 2,3-Dihydrothieno[3,4-b]-1,4-dioxine-5,7-dicarboxylic Acid

Formula: C₈H₆O₆S
MDL Number: MFCD00966477
CAS Number: 18361-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed099e0ca2386988be406cca470b533f26feb220&vid=ed099e0ca2386988be406cca470b533f26feb220&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR