2-Ethoxythiazole ≥98.0% (by GC)

Supplier: TCI America

E0669-25G E0669-5G
TCE0669-25GEA 105.86 CAD
TCE0669-25G TCE0669-5G
2-Ethoxythiazole ≥98.0% (by GC)
2-Ethoxythiazole

CAS Number: 15679-19-3
Molecular Formula: C5H7NOS
Molecular Weight: 129.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Specific Gravity (20/20): 1.14

Formula: C₅H₇NOS
Boiling Pt: 160 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00055026
CAS Number: 15679-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84936bad4b9aae058ae6670818f59f5cd2f1d49a&vid=84936bad4b9aae058ae6670818f59f5cd2f1d49a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR