4-Ethynylanisole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene

E0603-1G E0603-5G E0603-25G
TCE0603-1GEA 96.38 CAD
TCE0603-1G TCE0603-5G TCE0603-25G
4-Ethynylanisole ≥98.0% (by GC)
4-Ethynylanisole

CAS Number: 768-60-5
MDL Number: MFCD00168815
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 86
Flash Point (°C): 82
Freezing point (°C): 26
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 1-Ethynyl-4-methoxybenzene

Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 86 °C
Flash Pt: 82 °C
MDL Number: MFCD00168815
CAS Number: 768-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748af9f24c39b92c2965ae39406fe2a2adb4ac40&vid=748af9f24c39b92c2965ae39406fe2a2adb4ac40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR