1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide, [EMIIm], 1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide, 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulphonyl)imide, 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide
CAS Number: 174899-82-2
MDL Number: MFCD03788927
Molecular Formula: C8H11F6N3O4S2
Molecular Weight: 391.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Melting point (°C): -15
Specific Gravity (20/20): 1.53
Formula:
C₈H₁₁F₆N₃O₄S₂ MW: 391.32 g/mol Melting Pt: –15 °C Density: 1.53 (20 °C) |
MDL Number:
MFCD03788927 CAS Number: 174899-82-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...