1-Ethyl-3-methylimidazoliumhexafluorophosphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

E0493-25G E0493-5G
TCE0493-25GEA 469.26 CAD
TCE0493-25G TCE0493-5G
1-Ethyl-3-methylimidazoliumhexafluorophosphate ≥98.0% (by titrimetric analysis)
1-Ethyl-3-methylimidazolium hexafluorophosphate

[for Molten Salt]
CAS Number: 155371-19-0
MDL Number: MFCD00216703
Molecular Formula: C6H11F6N2P
Molecular Weight: 256.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 61

Formula: C₆H₁₁F₆N₂P
MW: 256.13 g/mol
Melting Pt: 61 °C
MDL Number: MFCD00216703
CAS Number: 155371-19-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55cb29da5f67f6891d0428020348cf9fe5b8a21&vid=77f9ed30cb605bd5bd9436a4d20c143eb545de9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR