3-Ethoxy-2-nitropyridine ≥95.0% (by GC)

Supplier: TCI America

E0390-1G E0390-5G
TCE0390-1GEA 105.86 CAD
TCE0390-1G TCE0390-5G
3-Ethoxy-2-nitropyridine ≥95.0% (by GC)
3-Ethoxy-2-nitropyridine

CAS Number: 74037-50-6
MDL Number: MFCD00006257
Molecular Formula: C7H8N2O3
Molecular Weight: 168.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 175
Freezing point (°C): 25

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Boiling Pt: 175 °C
MDL Number: MFCD00006257
CAS Number: 74037-50-6
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9861390210413a2778b13d7f52287fa64fd85bd&vid=a9861390210413a2778b13d7f52287fa64fd85bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR