Decyltrimethoxysilane ≥97.0%

Supplier: TCI America

D4704-250G D4704-25G
TCD4704-250GEA 431.34 CAD
TCD4704-250G TCD4704-25G
Decyltrimethoxysilane ≥97.0%
Decyltrimethoxysilane

CAS Number: 5575-48-4
MDL Number: MFCD00069135
Molecular Formula: C13H30O3Si
Molecular Weight: 262.47
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 117
Specific Gravity (20/20): 0.90

Formula: C₁₃H₃₀O₃Si
MDL Number: MFCD00069135
CAS Number: 5575-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95354114669b5f4c0871b4a262ad03997da40fee&vid=95354114669b5f4c0871b4a262ad03997da40fee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR