1,4-Diphenyl-2-butyne-1,4-dione ≥96.0%

Supplier: TCI America

D4699-1G D4699-250MG
TCD4699-1GEA 649.38 CAD
TCD4699-1G TCD4699-250MG
1,4-Diphenyl-2-butyne-1,4-dione ≥96.0%
1,4-Diphenyl-2-butyne-1,4-dione

CAS Number: 1087-09-8
Molecular Formula: C16H10O2
Molecular Weight: 234.25
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Diketone Compounds (Multi-component Reaction)
Other Product Names / Synonyms: Dibenzoylacetylene

Formula: C₁₆H₁₀O₂
Melting Pt: 113 °C
CAS Number: 1087-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1b0e71419700172ba00911185b194cdcd755906&vid=d1b0e71419700172ba00911185b194cdcd755906&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR