2,6-Dimethoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D4690-1G
D4690-5G
TCD4690-1GEA
137.46
CAD
TCD4690-1G
TCD4690-5G
2,6-Dimethoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
2,6-Dimethoxy-3-pyridineboronic acid
CAS Number: 221006-70-8
MDL Number: MFCD03788239
Molecular Formula: C7H10BNO4
Molecular Weight: 182.97
Form: Crystal
Color: White
Melting point (°C): 109
Other Product Names / Synonyms: 2,6-Dimethoxy-3-pyridylboronic Acid
Formula:
C₇H₁₀BNO₄ Melting Pt: 109 °C |
MDL Number:
MFCD03788239 CAS Number: 221006-70-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...