Diethylene glycol butyl methyl ether ≥99.0%

Supplier: TCI America

Synonyms: 1-[2-(2-Methoxyethoxy)ethoxy]butane

D4676-25ML D4676-500ML
TCD4676-25MLEA 56.88 CAD
TCD4676-25ML TCD4676-500ML
Diethylene glycol butyl methyl ether ≥99.0%
Diethylene glycol butyl methyl ether

CAS Number: 7382-32-3
MDL Number: MFCD00518661
Molecular Formula: C9H20O3
Molecular Weight: 176.26
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 1-[2-(2-Methoxyethoxy)ethoxy]butane

Formula: C₉H₂₀O₃
MW: 176.26 g/mol
Density: 0.91 (20 °C)
MDL Number: MFCD00518661
CAS Number: 7382-32-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe9b7938734dff8f066bae32a2ac186c8c39c96b&vid=fe9b7938734dff8f066bae32a2ac186c8c39c96b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR