2,6-Di(1-pyrazolyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

D4672-1G D4672-5G
TCD4672-1GEA 252.96 CAD
TCD4672-1G TCD4672-5G
2,6-Di(1-pyrazolyl)pyridine ≥98.0% (by GC)
2,6-Di(1-pyrazolyl)pyridine

CAS Number: 123640-38-0
Molecular Formula: C11H9N5
Molecular Weight: 211.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139

Formula: C₁₁H₉N₅
Melting Pt: 139 °C
MDL Number: MFCD28336856
CAS Number: 123640-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f7117ac603156759f95fb23712e6acebf8b1cc&vid=e9f7117ac603156759f95fb23712e6acebf8b1cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR