3,5-Dibromophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D4670-25G
D4670-5G
TCD4670-25GEA
276.03
CAD
TCD4670-25G
TCD4670-5G
3,5-Dibromophenylboronic acid (contains varying amounts of Anhydride)
3,5-Dibromophenylboronic acid
CAS Number: 117695-55-3
MDL Number: MFCD01075725
Molecular Formula: C6H5BBr2O2
Molecular Weight: 279.72
Form: Crystal
Color: Very Pale Yellow
Other Product Names / Synonyms: 3,5-Dibromobenzeneboronic Acid
Formula:
C₆H₅BBr₂O₂ MW: 279.72 g/mol |
MDL Number:
MFCD01075725 CAS Number: 117695-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...