1,5-Dibromonaphthalene ≥98.0% (by GC)

Supplier: TCI America

D4660-1G D4660-5G
TCD4660-1GEA 271.13 CAD
TCD4660-1G TCD4660-5G
1,5-Dibromonaphthalene ≥98.0% (by GC)
1,5-Dibromonaphthalene

CAS Number: 7351-74-8
MDL Number: MFCD00089984
Molecular Formula: C10H6Br2
Molecular Weight: 285.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 326
Melting point (°C): 130

Formula: C₁₀H₆Br₂
Boiling Pt: 326 °C
Melting Pt: 130 °C
MDL Number: MFCD00089984
CAS Number: 7351-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05ebc1880f849526e394a6e9daf05ff96757d1a7&vid=05ebc1880f849526e394a6e9daf05ff96757d1a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR