1,3-Dimethylimidazolium iodide ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dimethyl-1H-imidazol-3-ium iodide

D4654-25G D4654-5G
TCD4654-25GEA 580.22 CAD
TCD4654-25G TCD4654-5G
1,3-Dimethylimidazolium iodide ≥98.0%
1,3-Dimethylimidazolium iodide

CAS Number: 4333-62-4
MDL Number: MFCD00159685
Molecular Formula: C5H9IN2
Molecular Weight: 224.05
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 86
Lambda max.: 226 nm

Formula: C₅H₉IN₂
MW: 224.04 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00159685
CAS Number: 4333-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e8009bba4536e7adb39b0fb79d3bf83b41063c0&vid=6e8009bba4536e7adb39b0fb79d3bf83b41063c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR