2,6-Dichloro-3-formylpyridine ≥98.0%
Supplier: TCI America
Synonyms:
2,6-Dichloronicotinaldehyde, 2,6-Dichloropyridine-3-carboxaldehyde, 2,6-Dichloro-3-formylpyridine, 2,6-Dichloro-3-pyridinecarboxaldehyde
D4651-1G
D4651-5G
TCD4651-1GEA
82.16
CAD
TCD4651-1G
TCD4651-5G
2,6-Dichloro-3-formylpyridine ≥98.0%
2,6-Dichloro-3-formylpyridine
CAS Number: 55304-73-9
MDL Number: MFCD07369746
Molecular Formula: C6H3Cl2NO
Molecular Weight: 176.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 79
Other Product Names / Synonyms: 2,6-Dichloronicotinaldehyde, 2,6-Dichloro-3-formylpyridine
Formula:
C₆H₃Cl₂NO Boiling Pt: 95 °C Melting Pt: 79 °C |
MDL Number:
MFCD07369746 CAS Number: 55304-73-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...