Diethylamine hydroiodide ≥97.0%

Supplier: TCI America

D4643-1G D4643-5G
TCD4643-1GEA 72.68 CAD
TCD4643-1G TCD4643-5G
Diethylamine hydroiodide ≥97.0%
Diethylamine hydroiodide

CAS Number: 19833-78-4
Molecular Formula: C4H11N
Molecular Weight: 201.05
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Melting point (°C): 175

Formula: C₄H₁₁N
MW: 201.05 g/mol
Melting Pt: 175 °C
CAS Number: 19833-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=301f89c75984b6e78d859215cbe3958977c7878f&vid=301f89c75984b6e78d859215cbe3958977c7878f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR