3',5'-Dibromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D4637-25G
D4637-5G
TCD4637-25GEA
211.56
CAD
TCD4637-25G
TCD4637-5G
3',5'-Dibromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
3',5'-Dibromo-2'-hydroxyacetophenone
CAS Number: 22362-66-9
MDL Number: MFCD00191992
Molecular Formula: C8H6Br2O2
Molecular Weight: 293.94
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 109
Formula:
C₈H₆Br₂O₂ Melting Pt: 109 °C |
MDL Number:
MFCD00191992 CAS Number: 22362-66-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...