Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dimethoxycarbonyl-2,2'-bipyridine, 2,2'-Bipyridine-4,4'-dicarboxylic acid dimethyl ester, Dimethyl 2,2'-Bipyridine-4,4'-dicarboxylate, 4,4'-Bis(methoxycarbonyl)-2,2'-bipyridine

D4635-1G D4635-5G
TCD4635-1GEA 161.16 CAD
TCD4635-1G TCD4635-5G
Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate ≥98.0%
Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate

CAS Number: 71071-46-0
MDL Number: MFCD02669916
Molecular Formula: C14H12N2O4
Molecular Weight: 272.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 210


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Solar Cell Research Reagents
Other Product Names / Synonyms: 2,2'-Bipyridine-4,4'-dicarboxylic Acid Dimethyl Ester, 4,4'-Bis(methoxycarbonyl)-2,2'-bipyridine, 4,4'-Dimethoxycarbonyl-2,2'-bipyridine

Formula: C₁₄H₁₂N₂O₄
MW: 272.2600 g/mol
Melting Pt: 210 °C
MDL Number: MFCD02669916
CAS Number: 71071-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa54ba0637acf56dd73462489eee9fbcdbb2b77&vid=5aa54ba0637acf56dd73462489eee9fbcdbb2b77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR