Dimethyl-bis-(phenylethynyl)silane ≥98.0% (by GC)

Supplier: TCI America

D4625-1G D4625-5G
TCD4625-1GEA 140.62 CAD
TCD4625-1G TCD4625-5G
Dimethyl-bis-(phenylethynyl)silane ≥98.0% (by GC)
Dimethyl-bis-(phenylethynyl)silane

CAS Number: 2170-08-3
MDL Number: MFCD00270967
Molecular Formula: C18H16Si
Molecular Weight: 260.41
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 81
Lambda max.: 249 nm (Cyclohexane)

Formula: C₁₈H₁₆Si
Boiling Pt: 177 °C
Melting Pt: 81 °C
MDL Number: MFCD00270967
CAS Number: 2170-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d7bf82fe7fbd03e6a85a4d464eb34a489baa7f&vid=40d7bf82fe7fbd03e6a85a4d464eb34a489baa7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR