2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
Supplier: TCI America
D4623-1G
D4623-5G
TCD4623-1GEA
71.1
CAD
TCD4623-1G
TCD4623-5G
2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
2,3-Difluoro-5-(trifluoromethyl)pyridine
CAS Number: 89402-42-6
MDL Number: MFCD04972837
Molecular Formula: C6H2F5N
Molecular Weight: 183.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 100
Specific Gravity (20/20): 1.49
Formula:
C₆H₂F₅N Boiling Pt: 104 °C Density: 1.49 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD04972837 CAS Number: 89402-42-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...