2,3-Difluorobenzyl alcohol ≥98.0%

Supplier: TCI America

D4619-25G D4619-5G
TCD4619-25GEA 350.76 CAD
TCD4619-25G TCD4619-5G
2,3-Difluorobenzyl alcohol ≥98.0%

CAS Number: 75853-18-8
MDL Number: MFCD00010312
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 94
Specific Gravity (20/20): 1.30

Formula: C₇H₆F₂O
Density: 1.30 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00010312
CAS Number: 75853-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c75815a533a51fabfb2f7ef06ae9623a16bf5c3e&vid=c75815a533a51fabfb2f7ef06ae9623a16bf5c3e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR