Diethyl-2-propynylmalonate ≥97.0% (by GC)

Supplier: TCI America

D4616-1G D4616-5G
TCD4616-1GEA 246.32 CAD
TCD4616-1G TCD4616-5G
Diethyl-2-propynylmalonate ≥97.0% (by GC)
Diethyl-2-propynylmalonate

CAS Number: 17920-23-9
MDL Number: MFCD00091967
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.05


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 2-Propynylmalonic Acid Diethyl Ester, Diethyl Propargylmalonate, Propargylmalonic Acid Diethyl Ester

Formula: C₁₀H₁₄O₄
Boiling Pt: 115 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00091967
CAS Number: 17920-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1c5f181e556ce9527ff0d78baa5a868cfbe2cb6&vid=a1c5f181e556ce9527ff0d78baa5a868cfbe2cb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR