9,9-Dimethyl-2-nitrofluorene ≥98.0%

Supplier: TCI America

Synonyms: 9,9-Dimethyl-2-nitro-9H-fluorene

D4609-25G D4609-5G
TCD4609-25GEA 268.6 CAD
TCD4609-25G TCD4609-5G
9,9-Dimethyl-2-nitrofluorene ≥98.0%
9,9-Dimethyl-2-nitrofluorene

CAS Number: 605644-46-0
MDL Number: MFCD17170053
Molecular Formula: C15H13NO2
Molecular Weight: 239.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103
Lambda max.: 336 nm (CHCl3)

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
Melting Pt: 103 °C
MDL Number: MFCD17170053
CAS Number: 605644-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc2997ce507bb46dc6ac34770d3b4fd7270c4f7c&vid=cc2997ce507bb46dc6ac34770d3b4fd7270c4f7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR