4,4'-Diiodo-2,2'-dimethylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Diiodo-2,2'-dimethyl-1,1'-biphenyl

D4608-1G D4608-5G
TCD4608-1GEA 154.84 CAD
TCD4608-1G TCD4608-5G
4,4'-Diiodo-2,2'-dimethylbiphenyl ≥98.0%
4,4'-Diiodo-2,2'-dimethylbiphenyl

CAS Number: 69571-02-4
MDL Number: MFCD13185944
Molecular Formula: C14H12I2
Molecular Weight: 434.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98

Formula: C₁₄H₁₂I₂
MW: 434.06 g/mol
Melting Pt: 98 °C
MDL Number: MFCD13185944
CAS Number: 69571-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1644f20ececace24e443c80d96cb2e8979775d7f&vid=1644f20ececace24e443c80d96cb2e8979775d7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR