Diethyl (trichloromethyl)phosphonate ≥96.0%

Supplier: TCI America

Synonyms: Diethyltrichloromethylphosphonate, (Trichloromethyl)phosphonic Acid Diethyl Ester

D4607-25G D4607-5G
TCD4607-25GEA 643.06 CAD
TCD4607-25G TCD4607-5G
Diethyl (trichloromethyl)phosphonate ≥96.0%
Diethyl (trichloromethyl)phosphonate

CAS Number: 866-23-9
MDL Number: MFCD00013666
Molecular Formula: C5H10Cl3O3P
Molecular Weight: 255.46
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 113
Specific Gravity (20/20): 1.37
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Carbon Homologation / Degradation Reaction, Click Chemistry
Other Product Names / Synonyms: (Trichloromethyl)phosphonic Acid Diethyl Ester

Formula: C₅H₁₀Cl₃O₃P
MW: 255.46 g/mol
Boiling Pt: 135 °C
Density: 1.37 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00013666
CAS Number: 866-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f117ea4216f1c26e2ef33d84d0a8e12eb726cd1f&vid=f117ea4216f1c26e2ef33d84d0a8e12eb726cd1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR