2,3-Dibromonaphthalene ≥98.0%

Supplier: TCI America

Synonyms: 2,3-Diiodonaphthalene

D4597-1G D4597-200MG
TCD4597-1GEA 204.33 CAD
TCD4597-1G TCD4597-200MG
2,3-Dibromonaphthalene ≥98.0%
2,3-Dibromonaphthalene

CAS Number: 13214-70-5
MDL Number: MFCD00021589
Molecular Formula: C10H6Br2
Molecular Weight: 285.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 142
Lambda max.: 286 nm

Formula: C₁₀H₆Br₂
MW: 285.97
Melting Pt: 142 °C
MDL Number: MFCD00021589
CAS Number: 13214-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2820604657df0f6dcdaab90b0d8b19fe2d8d045b&vid=2820604657df0f6dcdaab90b0d8b19fe2d8d045b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR