(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate ≥98.0%
Supplier: TCI America
Synonyms:
1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-α-D-arabinofuranose, 2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-α-D-arabinofuranose
D4594-1G
D4594-5G
TCD4594-1GEA
59.57
CAD
TCD4594-1G
TCD4594-5G
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate ≥98.0%
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate
CAS Number: 97614-43-2
MDL Number: MFCD00083339
Molecular Formula: C26H21FO7
Molecular Weight: 464.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85
Formula:
C₂₆H₂₁FO₇ MW: 464.44 g/mol Melting Pt: 85 °C |
MDL Number:
MFCD00083339 CAS Number: 97614-43-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...