Diethyl (chloromethyl)phosphonate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(Chloromethyl)phosphonic Acid Diethyl Ester
CAS Number: 3167-63-3
MDL Number: MFCD00010189
Molecular Formula: C5H12ClO3P
Molecular Weight: 186.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 86
Specific Gravity (20/20): 1.20
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Nucleophiles (Aromatic VNS Reaction)
Other Product Names / Synonyms: (Chloromethyl)phosphonic Acid Diethyl Ester
Formula:
C₅H₁₂ClO₃P MW: 186.57 g/mol Boiling Pt: 110 °C Density: 1.20 (20 °C) Flash Pt: 86 °C |
MDL Number:
MFCD00010189 CAS Number: 3167-63-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...