1,3-Dibromo-2,2-dimethoxypropane ≥98.0% (by GC)
Supplier: TCI America
D4579-25G
D4579-5G
TCD4579-25GEA
406.06
CAD
TCD4579-25G
TCD4579-5G
1,3-Dibromo-2,2-dimethoxypropane ≥98.0% (by GC)
1,3-Dibromo-2,2-dimethoxypropane
CAS Number: 22094-18-4
MDL Number: MFCD00051792
Molecular Formula: C5H10Br2O2
Molecular Weight: 261.94
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65
Other Product Names / Synonyms: 1,3-Dibromoacetone Dimethyl Acetal
Formula:
BrH₂CC(OCH₃)₂CH₂Br MW: 261.94 g/mol Melting Pt: 65 °C |
MDL Number:
MFCD00051792 CAS Number: 22094-18-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...