(R)-1-(2,6-Dichloro-3-fluorophenyl)ethanol ≥98.0% (by GC)
Supplier: TCI America
D4577-1G
D4577-5G
TCD4577-1GEA
36.34
CAD
TCD4577-1G
TCD4577-5G
(R)-1-(2,6-Dichloro-3-fluorophenyl)ethanol ≥98.0% (by GC)
(R)-1-(2,6-Dichloro-3-fluorophenyl)ethanol
CAS Number: 330156-50-8
MDL Number: MFCD09863794
Molecular Formula: C8H7Cl2FO
Molecular Weight: 209.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 43
Specific rotation [a]20/D: -8.0 deg (C=5, MeOH)
Formula:
C₈H₇Cl₂FO Melting Pt: 43 °C |
MDL Number:
MFCD09863794 CAS Number: 330156-50-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...