3,5-Dichlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D4575-1G D4575-5G
TCD4575-1GEA 63.2 CAD
TCD4575-1G TCD4575-5G
3,5-Dichlorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Dichlorophenylboronic acid pinacol ester

CAS Number: 68716-51-8
MDL Number: MFCD05863906
Molecular Formula: C12H15BCl2O2
Molecular Weight: 272.96
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 53


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 1,3-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dichlorophenylboronic Acid Pinacol Ester

Formula: C₁₂H₁₅BCl₂O₂
Melting Pt: 53 °C
MDL Number: MFCD05863906
CAS Number: 68716-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=242932ffcdd51ac563f31ff33c6bd3c46fb0edf0&vid=242932ffcdd51ac563f31ff33c6bd3c46fb0edf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR