2,5-Dibromo-3-decylthiophene ≥97.0% (by GC)

Supplier: TCI America

D4572-25G D4572-5G
TCD4572-25GEA 285.98 CAD
TCD4572-25G TCD4572-5G
2,5-Dibromo-3-decylthiophene ≥97.0% (by GC)
2,5-Dibromo-3-decylthiophene

CAS Number: 158956-23-1
MDL Number: MFCD00274314
Molecular Formula: C14H22Br2S
Molecular Weight: 382.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 113
Specific Gravity (20/20): 1.37
Storage Temperature: 0-10°C

Formula: C₁₄H₂₂Br₂S
MW: 382.20 g/mol
Boiling Pt: 148 °C
Density: 1.37 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00274314
CAS Number: 158956-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=259ec1d8093f25d41329a1019843da22e9aa7f3f&vid=259ec1d8093f25d41329a1019843da22e9aa7f3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR