4-(2,6-Dichloro-4-pyrimidyl)morpholine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-(2,6-Dichloropyrimidin-4-yl)morpholine, 2,4-Dichloro-6-(4-morpholino)pyrimidine

D4571-1G D4571-5G
TCD4571-1GEA 218.2 CAD
TCD4571-1G TCD4571-5G
4-(2,6-Dichloro-4-pyrimidyl)morpholine ≥98.0% (by GC)
4-(2,6-Dichloro-4-pyrimidyl)morpholine

CAS Number: 52127-83-0
MDL Number: MFCD09746259
Molecular Formula: C8H9Cl2N3O
Molecular Weight: 234.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 2,4-Dichloro-6-(4-morpholino)pyrimidine

Formula: C₈H₉Cl₂N₃O
MW: 234.08 g/mol
Melting Pt: 125 °C
MDL Number: MFCD09746259
CAS Number: 52127-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ddd3823a65dec8b17e252d3b895a653c18678ac&vid=6ddd3823a65dec8b17e252d3b895a653c18678ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR