2,7-Dibromo-9,9-dipropylfluorene ≥98.0%
Supplier: TCI America
D4569-1G
D4569-5G
TCD4569-1GEA
143.78
CAD
TCD4569-1G
TCD4569-5G
2,7-Dibromo-9,9-dipropylfluorene ≥98.0%
2,7-Dibromo-9,9-dipropylfluorene
CAS Number: 157771-56-7
MDL Number: MFCD12024280
Molecular Formula: C19H20Br2
Molecular Weight: 408.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139
Formula:
C₁₉H₂₀Br₂ MW: 408.18 g/mol Melting Pt: 139 °C |
MDL Number:
MFCD12024280 CAS Number: 157771-56-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...