3,6-Dibromo-9-(4-bromophenyl)carbazole ≥98.0%
Supplier: TCI America
Synonyms:
3,6-Dibromo-9-(4-bromophenyl)-9H-carbazole
D4563-1G
D4563-200MG
TCD4563-1GEA
638
CAD
TCD4563-1G
TCD4563-200MG
3,6-Dibromo-9-(4-bromophenyl)carbazole ≥98.0%
3,6-Dibromo-9-(4-bromophenyl)carbazole
CAS Number: 73087-83-9
MDL Number: MFCD12024284
Molecular Formula: C18H10Br3N
Molecular Weight: 480.00
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 209
Lambda max.: 304 nm (CHCl3)
Formula:
C₁₈H₁₀Br₃N Melting Pt: 209 °C |
MDL Number:
MFCD12024284 CAS Number: 73087-83-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...