2,7-Dibromo-9,9-didecylfluorene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4561-25G D4561-5G
TCD4561-25GEA 338.91 CAD
TCD4561-25G TCD4561-5G
2,7-Dibromo-9,9-didecylfluorene ≥98.0% (by HPLC, titration analysis)
2,7-Dibromo-9,9-didecylfluorene

CAS Number: 175922-78-8
MDL Number: MFCD00971288
Molecular Formula: C33H48Br2
Molecular Weight: 604.56
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 40

Formula: C₃₃H₄₈Br₂
Melting Pt: 40 °C
MDL Number: MFCD00971288
CAS Number: 175922-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66efa01c38145331eb700ab4706b8b4693d3dfa6&vid=66efa01c38145331eb700ab4706b8b4693d3dfa6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR