Dimethylamine hydroiodide ≥98.0%

Supplier: TCI America

D4555-1G D4555-5G
TCD4555-1GEA 94.17 CAD
TCD4555-1G TCD4555-5G
Dimethylamine hydroiodide ≥98.0%
Dimethylamine hydroiodide

CAS Number: 51066-74-1
Molecular Formula: C2H7N
Molecular Weight: 173.00
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 155

Formula: C₂H₇N
MW: 173.00 g/mol
Melting Pt: 155 °C
CAS Number: 51066-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f57606d4f7a2411607f5d735dfb77ac98e507ae&vid=9f57606d4f7a2411607f5d735dfb77ac98e507ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR